2-Chloro-6-iodopyridine 98%

Supplier: Apollo Scientific

OR8714-5G
APOSOR8714-5GEA 101 EUR
APOSOR8714-5G
2-Chloro-6-iodopyridine 98%
2-Chloro-6-iodopyridine
Formula: C₅H₃ClIN
Storage Temperature: Ambient
MDL Number: MFCD07784182
CAS Number: 258506-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR