(R)-(-)-4-Phenyl-2-oxazolidinone 97%

Supplier: Apollo Scientific
Warning

Synonyms: (R)-4-Phenyloxazolidin-2-one

OR8746-100G OR8746-25G OR8746-500G
APOSOR8746-100GEA 31.9 EUR
APOSOR8746-100G APOSOR8746-25G APOSOR8746-500G
(R)-(-)-4-Phenyl-2-oxazolidinone 97%
(R)-(-)-4-Phenyl-2-oxazolidinone
Formula: C₉H₉NO₂
MW: 163,18 g/mol
Melting Pt: 129…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00192393
CAS Number: 90319-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR