4-(Morpholinomethyl)phenylboronic acid pinacol ester

Supplier: Apollo Scientific
Warning

Synonyms: 4-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester, 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]morpholine, 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine, 4,4,5,5-Tetramethyl-2-[4-(morpholinomethyl)phenyl]-1,3,2-dioxaborolane, 4-(Morpholinomethyl)phenylboronic acid Pinacol ester

OR8756-1G OR8756-5G
APOSOR8756-1GEA 53.9 EUR
APOSOR8756-1G APOSOR8756-5G
4-(Morpholinomethyl)phenylboronic acid pinacol ester
4-(Morpholinomethyl)phenylboronic acid pinacol ester
Formula: C₁₇H₂₆BNO₃
MW: 303,20 g/mol
Melting Pt: 84…89 °C
Storage Temperature: Ambient
MDL Number: MFCD04974052
CAS Number: 364794-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR