4-(4-Methoxyphenyl)thiophene-2-carboxylic acid 95+%

Supplier: Apollo Scientific
Warning

OR8778-1G OR8778-5G
APOSOR8778-1GEA 319 EUR
APOSOR8778-1G APOSOR8778-5G
4-(4-Methoxyphenyl)thiophene-2-carboxylic acid 95+%
4-(4-Methoxyphenyl)thiophene-2-carboxylic acid
Formula: C₁₂H₁₀O₃S
MW: 234.28 g/mol
Melting Pt: 193…169 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00203285
CAS Number: 82437-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR