Methyl-4-piperidinecarboxylate 97%
Supplier: Apollo Scientific
Synonyms:
Methyl 4-Piperidinecarboxylate, Methyl isonipecotate, Methyl piperidine-4-carboxylate
OR8795-500G
APOSOR8795-500GEA
186
EUR
APOSOR8795-500G
Methyl-4-piperidinecarboxylate 97%
Methyl 4-piperidinecarboxylate
Formula:
C₇H₁₃NO₂ MW: 143,18 g/mol Boiling Pt: 85…90 °C Density: 1,06 g/cm³ (25 °C) Flash Pt: 89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190578 CAS Number: 2971-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...