2-(4-Pyridyl)ethanesulphonic acid

Supplier: Apollo Scientific
Danger

OR8834-25G OR8834-5G OR8834-100G
APOSOR8834-25GEA 66.7 EUR
APOSOR8834-25G APOSOR8834-5G APOSOR8834-100G
2-(4-Pyridyl)ethanesulphonic acid
2-(4-Pyridyl)ethanesulphonic acid
Formula: C₇H₉NO₃S
MW: 187,21 g/mol
Melting Pt: 295…299 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006452
CAS Number: 53054-76-5
EINECS: 258-331-3
UN: 2585
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR