2-Bromo-3-chloropyridine

Supplier: Apollo Scientific
Warning

OR8849-5G OR8849-25G OR8849-100G
APOSOR8849-5GEA 14.7 EUR
APOSOR8849-5G APOSOR8849-25G APOSOR8849-100G
2-Bromo-3-chloropyridine
2-Bromo-3-chloropyridine
Formula: C₅H₃BrClN
Melting Pt: 58…63 °C
Storage Temperature: Ambient
MDL Number: MFCD04039344
CAS Number: 96424-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR