4-Iodotoluene 98%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Iodo-4-methylbenzene

OR8875-100G OR8875-25G OR8875-500G
APOSOR8875-100GEA 43.5 EUR
APOSOR8875-100G APOSOR8875-25G APOSOR8875-500G
4-Iodotoluene 98%
4-Iodotoluene
Formula: C₇H₇I
MW: 218.04 g/mol
Boiling Pt: 210…212 °C
Melting Pt: 33…35 °C
Density: 1.678 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00001059
CAS Number: 624-31-7
EINECS: 210-841-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR