2,6-Lutidine-N-oxide

Supplier: Apollo Scientific
Warning

Synonyms: 2,6-Dimethylpyridine N-Oxide

OR8899-250G
APOSOR8899-250GEA 698 EUR
APOSOR8899-250G
2,6-Lutidine-N-oxide
2,6-Lutidine-N-oxide
Formula: C₇H₉NO
MW: 123,16 g/mol
Boiling Pt: 108…111 °C/10mmHg
Density: 1,102 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039715
CAS Number: 1073-23-0
EINECS: 214-025-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR