3-Methyl-4-nitro-1H-pyrazole 97%
Supplier: Apollo Scientific
|
Warning
|
OR8929-25G
OR8929-100G
OR8929-500G
APOSOR8929-25GEA
21.9
EUR
APOSOR8929-25G
APOSOR8929-100G
APOSOR8929-500G
3-Methyl-4-nitro-1H-pyrazole 97%
3-Methyl-4-nitro-1H-pyrazole
Formula:
C4H5N3O2 Boiling Pt: 325 °C Melting Pt: 131…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037864 CAS Number: 5334-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...