4-(Methylsulphonyl)phenylacetic acid 95+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(4-(Methylsulphonyl)phenyl)acetic acid
OR8936-1G
OR8936-5G
OR8936-25G
APOSOR8936-1GEA
49.5
EUR
APOSOR8936-1G
APOSOR8936-5G
APOSOR8936-25G
4-(Methylsulphonyl)phenylacetic acid 95+%
4-(Methylsulphonyl)phenylacetic acid
Formula:
C₉H₁₀O₄S MW: 214,24 g/mol Melting Pt: 136…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216495 CAS Number: 90536-66-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...