(R)-(-)-Nipecotic acid

Supplier: Apollo Scientific
Warning

Synonyms: (R)-Piperidine-3-carboxylic acid, (R)-(-)-3-Piperidinecarboxylic acid, D-Nipecotic acid

OR8967-1G OR8967-25G OR8967-5G
APOSOR8967-1GEA 21.9 EUR
APOSOR8967-1G APOSOR8967-25G APOSOR8967-5G
(R)-(-)-Nipecotic acid
(R)-(-)-Nipecotic acid
Formula: C₆H₁₁NO₂
MW: 129,16 g/mol
Melting Pt: 250…255 °C
Storage Temperature: Ambient
MDL Number: MFCD01630787
CAS Number: 25137-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR