N,N-Diethylaniline
Supplier: Apollo Scientific
|
Danger
|
OR8987-1L
OR8987-500ML
APOSOR8987-1LEA
55.3
EUR
APOSOR8987-1L
APOSOR8987-500ML
N,N-Diethylaniline
N,N-Diethylaniline
Formula:
C₁₀H₁₅N MW: 149,24 g/mol Boiling Pt: 217 °C Melting Pt: –38 °C Density: 0,938 g/cm³ (25 °C) Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009042 CAS Number: 91-66-7 EINECS: 202-088-8 UN: 2432 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...