1,2,2,3-Tetrachloropropane 99%
Supplier: Apollo Scientific
OR8992-5G
OR8992-10G
APOSOR8992-5GEA
165
EUR
APOSOR8992-5G
APOSOR8992-10G
1,2,2,3-Tetrachloropropane 99%
1,2,2,3-Tetrachloropropane
Formula:
ClH₂CCCl₂CH₂Cl MW: 181,88 g/mol Boiling Pt: 87…88 °C/50mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00039362 CAS Number: 13116-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...