1-Benzylpyrrolidine-3-carbonitrile 96%
Supplier: Apollo Scientific
OR900130-5G
APOSOR900130-5GEA
58
EUR
APOSOR900130-5G
1-Benzylpyrrolidine-3-carbonitrile 96%
1-Benzylpyrrolidine-3-carbonitrile
Formula:
C₁₂H₁₄N₂ Storage Temperature: Ambient |
MDL Number:
MFCD05662780 CAS Number: 10603-52-8 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...