2-Cyclohepten-1-one 90%
Supplier: Apollo Scientific
|
Warning
|
OR900263-10G
OR900263-25G
APOSOR900263-10GEA
105
EUR
APOSOR900263-10G
APOSOR900263-25G
2-Cyclohepten-1-one 90%
2-Cyclohepten-1-one
Formula:
C₇H₁₀O Density: 0,988 g/cm³ (25 °C) Flash Pt: 68 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004157 CAS Number: 1121-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...