1,4-Phenylenediacetic acid 97%

Supplier: Apollo Scientific

Synonyms: 2,2'-(1,4-Phenylene)diacetic acid

OR900642-100G OR900642-25G
APOSOR900642-100GEA 261 EUR
APOSOR900642-100G APOSOR900642-25G
1,4-Phenylenediacetic acid 97%
1,4-Phenylenediacetic acid
Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004354
CAS Number: 7325-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR