4-Methoxy-m-toluidine 95%
Supplier: Apollo Scientific
Synonyms:
4-Methoxy-3-methylphenylamine, 4-Methoxy-3-methylaniline
OR900687-100G
OR900687-10G
OR900687-25G
APOSOR900687-100GEA
527
EUR
APOSOR900687-100G
APOSOR900687-10G
APOSOR900687-25G
4-Methoxy-m-toluidine 95%
4-Methoxy-m-toluidine
Formula:
C₈H₁₁NO MW: 137,1800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07779234 CAS Number: 136-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...