N,N-Dimethylacetamide dimethyl acetal 90%

Supplier: Apollo Scientific

Synonyms: (1,1-Dimethoxyethyl)dimethylamine, 1,1-Dimethoxy-N,N-dimethylethanamine

OR900738-100G
APOSOR900738-100GEA 88.6 EUR
APOSOR900738-100G
N,N-Dimethylacetamide dimethyl acetal 90%
N,N-Dimethylacetamide dimethyl acetal
Formula: C₆H₁₅NO₂
MW: 133,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008476
CAS Number: 18871-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR