(6-Bromo-3-pyridinyl)methanol 98%

Supplier: Apollo Scientific

Synonyms: 2-Bromo-5-(hydroxymethyl)pyridine, 2-Bromo-5-pyridinemethanol, (6-Bromopyridin-3-yl)methanol

OR900843-25G
APOSOR900843-25GEA 108 EUR
APOSOR900843-25G
(6-Bromo-3-pyridinyl)methanol 98%
(6-Bromo-3-pyridinyl)methanol
Formula: C₆H₆BrNO
MW: 188,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974508
CAS Number: 122306-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR