2-Chloro-8-methylquinoline 98%

Supplier: Apollo Scientific

OR901660-5G
APOSOR901660-5GEA 134 EUR
APOSOR901660-5G
2-Chloro-8-methylquinoline 98%
2-Chloro-8-methylquinoline
Formula: C₁₀H₈ClN
MW: 177,6300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11108682
CAS Number: 4225-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR