2-(5-Aminopyridin-2-yl)-2-methylpropanenitrile 95%

Supplier: Apollo Scientific

OR901829-100MG OR901829-1G OR901829-250MG
APOSOR901829-100MGEA 147 EUR
APOSOR901829-100MG APOSOR901829-1G APOSOR901829-250MG
2-(5-Aminopyridin-2-yl)-2-methylpropanenitrile 95%
2-(5-Aminopyridin-2-yl)-2-methylpropanenitrile
Formula: C₉H₁₁N₃
Storage Temperature: Ambient
CAS Number: 1226776-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR