5-Bromo-1-methylpyridin-2(1H)-one 98%
Supplier: Apollo Scientific
Synonyms:
5-Bromo-1-methyl-2(1H)-pyridinone
OR901973-5G
APOSOR901973-5GEA
60.9
EUR
APOSOR901973-5G
5-Bromo-1-methylpyridin-2(1H)-one 98%
5-Bromo-1-methylpyridin-2(1H)-one
Formula:
C₆H₆BrNO MW: 188,02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09972188 CAS Number: 81971-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...