Methyl-5-methylpyrazine-2-carboxylate 98%
Supplier: Apollo Scientific
Synonyms:
Methyl 5-methylpyrazine-2-carboxylate
OR902034-25G
APOSOR902034-25GEA
109
EUR
APOSOR902034-25G
Methyl-5-methylpyrazine-2-carboxylate 98%
Methyl-5-methylpyrazine-2-carboxylate
Formula:
C₇H₈N₂O₂ MW: 152,1500 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04116910 CAS Number: 41110-33-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...