8-Methoxyquinoline 98%

Supplier: Apollo Scientific

OR902049-10G OR902049-1G OR902049-5G
APOSOR902049-10GEA 317 EUR
APOSOR902049-10G APOSOR902049-1G APOSOR902049-5G
8-Methoxyquinoline 98%
8-Methoxyquinoline
Formula: C₁₀H₉NO
Storage Temperature: Ambient
MDL Number: MFCD00957068
CAS Number: 938-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR