2-[(2-Furylmethyl)amino]nicotinonitrile 98%

Supplier: Apollo Scientific

OR902138-1G OR902138-5G
APOSOR902138-1GEA 145 EUR
APOSOR902138-1G APOSOR902138-5G
2-[(2-Furylmethyl)amino]nicotinonitrile 98%
2-[(2-Furylmethyl)amino]nicotinonitrile
Formula: C₁₁H₉N₃O
MW: 199,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09455276
CAS Number: 945347-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR