(R)-Ethyl-2-hydroxypropanoate 95%
Supplier: Apollo Scientific
OR902275-100G
OR902275-25G
OR902275-5G
APOSOR902275-100GEA
53.8
EUR
APOSOR902275-100G
APOSOR902275-25G
APOSOR902275-5G
(R)-Ethyl-2-hydroxypropanoate 95%
(R)-Ethyl-2-hydroxypropanoate
Formula:
C₅H₁₀O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00077825 CAS Number: 7699-00-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...