Methyl-5-quinolyl ether 98%

Supplier: Apollo Scientific

Synonyms: 5-Methoxyquinoline

OR902277-10G OR902277-25G OR902277-5G
APOSOR902277-10GEA 227 EUR
APOSOR902277-10G APOSOR902277-25G APOSOR902277-5G
Methyl-5-quinolyl ether 98%
Methyl 5-quinolyl ether
Formula: C₁₀H₉NO
Storage Temperature: Ambient
MDL Number: MFCD02752664
CAS Number: 6931-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR