2-Benzylaniline 97%

Supplier: Apollo Scientific

Synonyms: 2-Aminodiphenylmethane, 2-Benzylaniline

OR902312-25G OR902312-5G
APOSOR902312-25GEA 256 EUR
APOSOR902312-25G APOSOR902312-5G
2-Benzylaniline 97%
2-Benzylaniline
Formula: C₁₃H₁₃N
MW: 183,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007750
CAS Number: 28059-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR