4-Amino-2-bromopyrimidine 97%

Supplier: Apollo Scientific
Warning

OR902755-1G OR902755-250MG OR902755-5G
APOSOR902755-1GEA 129 EUR
APOSOR902755-1G APOSOR902755-250MG APOSOR902755-5G
4-Amino-2-bromopyrimidine 97%
4-Amino-2-bromopyrimidine
Formula: C₄H₄BrN₃
MW: 174,0000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11109798
CAS Number: 1160994-71-7
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR