5-Bromo-1H-1,2,4-triazole-3-carboxylic acid 95%
Supplier: Apollo Scientific
OR902762-1G
OR902762-250MG
APOSOR902762-1GEA
139
EUR
APOSOR902762-1G
APOSOR902762-250MG
5-Bromo-1H-1,2,4-triazole-3-carboxylic acid 95%
5-Bromo-1H-1,2,4-triazole-3-carboxylic acid
Formula:
C₃H₂BrN₃O₂ MW: 191,97 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11040182 CAS Number: 674287-63-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...