Ethyl succinyl chloride 95%
Supplier: Apollo Scientific
Synonyms:
Ethyl 4-chloro-4-oxobutanoate
OR902965-100G
OR902965-25G
OR902965-500G
APOSOR902965-100GEA
190
EUR
APOSOR902965-100G
APOSOR902965-25G
APOSOR902965-500G
Ethyl succinyl chloride 95%
Ethyl succinyl chloride
Formula:
C₆H₉ClO₃ MW: 164,59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00000751 CAS Number: 14794-31-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...