Ethyl-5-chlorothiophene-2-carboxylate 97%

Supplier: Apollo Scientific

Synonyms: Ethyl 5-chlorothiophene-2-carboxylate

OR903157-25G
APOSOR903157-25GEA 120 EUR
APOSOR903157-25G
Ethyl-5-chlorothiophene-2-carboxylate 97%
Ethyl 5-chlorothiophene-2-carboxylate
Formula: C₇H₇ClO₂S
MW: 190,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051732
CAS Number: 5751-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR