5-Pentylbenzene-1,3-diol 98%
Supplier: Apollo Scientific
OR903168-1G
OR903168-25G
OR903168-5G
APOSOR903168-1GEA
21.9
EUR
APOSOR903168-1G
APOSOR903168-25G
APOSOR903168-5G
5-Pentylbenzene-1,3-diol 98%
5-Pentylbenzene-1,3-diol
Formula:
C₁₁H₁₆O₂ MW: 180,25 g/mol Boiling Pt: 162…164 °C Melting Pt: 46…48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002293 CAS Number: 500-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...