2,3-Dihydrobenzo[b][1,4]dioxin-5-ol 95%

Supplier: Apollo Scientific

OR903269-10G OR903269-1G OR903269-5G
APOSOR903269-10GEA 287 EUR
APOSOR903269-10G APOSOR903269-1G APOSOR903269-5G
2,3-Dihydrobenzo[b][1,4]dioxin-5-ol 95%
2,3-Dihydrobenzo[b][1,4]dioxin-5-ol
Formula: C₈H₈O₃
MW: 152,1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02103729
CAS Number: 10288-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR