Quinoxalin-5-ol 98%
Supplier: Apollo Scientific
Synonyms:
5-Hydroxyquinoxaline
OR903311-5G
APOSOR903311-5GEA
163
EUR
APOSOR903311-5G
Quinoxalin-5-ol 98%
Quinoxalin-5-ol
Formula:
C8H6N2O MW: 146,05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00972004 CAS Number: 17056-99-4 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...