Ethyl-6-chloro-3-pyridazinecarboxylate 98%

Supplier: Apollo Scientific

Synonyms: Ethyl 6-chloro-3-pyridazinecarboxylate, Ethyl 6-chloropyridazine-3-carboxylate, 6-chloro-3-pyridazinecarboxylic acid ethyl ester, 6-Chloro-pyridazine-3-carboxylic acid ethyl ester

OR903332-1G OR903332-5G
APOSOR903332-1GEA 93.1 EUR
APOSOR903332-1G APOSOR903332-5G
Ethyl-6-chloro-3-pyridazinecarboxylate 98%
Ethyl-6-chloro-3-pyridazinecarboxylate
Formula: C₇H₇ClN₂O₂
MW: 186,595 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09908168
CAS Number: 75680-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR