6-Chloropyrido[3,4-d]pyrimidin-4(3H)-one 98%

Supplier: Apollo Scientific

OR903355-1G
APOSOR903355-1GEA 62.5 EUR
APOSOR903355-1G
6-Chloropyrido[3,4-d]pyrimidin-4(3H)-one 98%
6-Chloropyrido[3,4-d]pyrimidin-4(3H)-one
Formula: C₇H₄ClN₃O
MW: 181,5800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044635
CAS Number: 171178-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR