3-Methyl-1H-indazol-7-ol 95%
Supplier: Apollo Scientific
OR903386-1G
OR903386-250MG
APOSOR903386-1GEA
865
EUR
APOSOR903386-1G
APOSOR903386-250MG
3-Methyl-1H-indazol-7-ol 95%
3-Methyl-1H-indazol-7-ol
Formula:
C8H8N2O Storage Temperature: Ambient |
CAS Number:
1131595-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...