3-Bromo-1-methyl-1H-pyrazole 98%
Supplier: Apollo Scientific
OR903550-10G
OR903550-1G
OR903550-5G
APOSOR903550-10GEA
118
EUR
APOSOR903550-10G
APOSOR903550-1G
APOSOR903550-5G
3-Bromo-1-methyl-1H-pyrazole 98%
3-Bromo-1-methyl-1H-pyrazole
Formula:
C4H5BrN2 MW: 161,0000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11110668 CAS Number: 151049-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...