5-Aminoimidazo[1,2-a]pyridine 95%

Supplier: Apollo Scientific

Synonyms: Imidazo[1,2-a]pyridin-5-amine

OR903639-5G
APOSOR903639-5GEA 68.3 EUR
APOSOR903639-5G
5-Aminoimidazo[1,2-a]pyridine 95%
5-Aminoimidazo[1,2-a]pyridine
Formula: C₇H₇N₃
MW: 133,1506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863377
CAS Number: 66358-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR