6-Ethoxypyridazin-3-amine 98%

Supplier: Apollo Scientific

OR903647-10G OR903647-1G OR903647-5G
APOSOR903647-10GEA 648 EUR
APOSOR903647-10G APOSOR903647-1G APOSOR903647-5G
6-Ethoxypyridazin-3-amine 98%
6-Ethoxypyridazin-3-amine
Formula: C₆H₉N₃O
MW: 139,1552 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460377
CAS Number: 39614-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR