2-Chloro-4-ethynylpyridine 98%

Supplier: Apollo Scientific

OR903659-5G
APOSOR903659-5GEA 255 EUR
APOSOR903659-5G
2-Chloro-4-ethynylpyridine 98%
2-Chloro-4-ethynylpyridine
Formula: C₇H₄ClN
MW: 137,5664 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189742
CAS Number: 945717-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR