1-(Benzo[d]oxazol-2-yl)ethanone 97%

Supplier: Apollo Scientific
Warning

OR903710-1G OR903710-5G
APOSOR903710-1GEA 183 EUR
APOSOR903710-1G APOSOR903710-5G
1-(Benzo[d]oxazol-2-yl)ethanone 97%
1-(Benzo[d]oxazol-2-yl)ethanone
Formula: C₉H₇NO₂
MW: 161,1574 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00223823
CAS Number: 122433-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR