[4-(2-Aminopropyl)phenyl]dimethylamine 95%

Supplier: Apollo Scientific

Synonyms: 4-(2-Aminopropyl)-n,n-dimethylaniline

OR903859-1G OR903859-250MG
APOSOR903859-1GEA 865 EUR
APOSOR903859-1G APOSOR903859-250MG
[4-(2-Aminopropyl)phenyl]dimethylamine 95%
[4-(2-Aminopropyl)phenyl]dimethylamine
Formula: C₁₁H₁₈N₂
MW: 178,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06662285
CAS Number: 57580-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR