6-Heptyn-1-ol 97%

Supplier: Apollo Scientific
Warning

OR903961-100G OR903961-10G OR903961-25G OR903961-5G
APOSOR903961-100GEA 408 EUR
APOSOR903961-100G APOSOR903961-10G APOSOR903961-25G APOSOR903961-5G
6-Heptyn-1-ol 97%
6-Heptyn-1-ol
Formula: C₇H₁₂O
MW: 112,17 g/mol
Boiling Pt: 105 °C
Density: 0,9 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD00049198
CAS Number: 63478-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR