2-Chloro-7-methylquinoxaline 95%
Supplier: Apollo Scientific
OR903996-1G
OR903996-250MG
APOSOR903996-1GEA
667
EUR
APOSOR903996-1G
APOSOR903996-250MG
2-Chloro-7-methylquinoxaline 95%
2-Chloro-7-methylquinoxaline
Formula:
C₉H₇ClN₂ Storage Temperature: Ambient |
CAS Number:
90272-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...