2-Naphthyl isocyanate 95%

Supplier: Apollo Scientific

OR904280-1G
APOSOR904280-1GEA 162 EUR
APOSOR904280-1G
2-Naphthyl isocyanate 95%
2-Naphthyl isocyanate

When melted, may contain small amount of sediment.

Formula: C₁₁H₇NO
MW: 169,18 g/mol
Boiling Pt: 83 °C
Melting Pt: 52…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00014319
CAS Number: 2243-54-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR