2-(2-Aminopyrimidin-5-yl)benzaldehyde 95%
Supplier: Apollo Scientific
OR904469-1G
OR904469-250MG
APOSOR904469-1GEA
84.1
EUR
APOSOR904469-1G
APOSOR904469-250MG
2-(2-Aminopyrimidin-5-yl)benzaldehyde 95%
2-(2-Aminopyrimidin-5-yl)benzaldehyde
Formula:
C₁₁H₉N₃O Storage Temperature: Ambient |
CAS Number:
914349-52-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...