1-(5-Methyl-2-pyridinyl)piperazine 97%
Supplier: Apollo Scientific
OR904676-10G
OR904676-1G
OR904676-5G
APOSOR904676-10GEA
899
EUR
APOSOR904676-10G
APOSOR904676-1G
APOSOR904676-5G
1-(5-Methyl-2-pyridinyl)piperazine 97%
1-(5-Methyl-2-pyridinyl)piperazine
Formula:
C₁₀H₁₅N₃ Storage Temperature: Ambient |
MDL Number:
MFCD08061050 CAS Number: 104395-86-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...